4-n-Butoxy-2,6-difluorobenzonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1373920-94-5
:
Formula:C₁₁H₁₁F₂NO
MDL Number:MFCD22201026
:211.21 g/mol
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
Frequently Bought Together