Order Entry
Italy
ContactUsLinkComponent
2,6-Difluoro-4-methoxybenzylamine 98%
Corrosive
2,6-Difluoro-4-methoxybenzylamine 98%
Catalog # APOSPC303024-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
2,6-Difluoro-4-methoxybenzylamine 98%
Catalog # APOSPC303024-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :173.16 g/mol
Storage Temperature:Ambient
UN:2735
ADR:8,III
 :GHS05
CAS Number:771573-20-7
Formula:C₈H₉F₂NO
MDL Number:MFCD04115924