
5-Chloro-2-(trifluoromethoxy)benzaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1092461-15-8
:
Formula:C₈H₄ClF₃O₂
MDL Number:MFCD11519353
:224.57 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together