4-Methoxy-3-(trifluoromethoxy)phenylacetic acid
Catalog # APOSPC302998-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:250.17 g/mol
Storage Temperature:Ambient
CAS Number:1261557-46-3
Formula:C₁₀H₉F₃O₄
MDL Number:MFCD18398521