Order Entry
Italy
ContactUsLinkComponent
4-Fluoro-2-(trifluoromethoxy)phenylacetic acid 98%
4-Fluoro-2-(trifluoromethoxy)phenylacetic acid 98%
Catalog # APOSPC302990-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Fluoro-2-(trifluoromethoxy)phenylacetic acid 98%
Catalog # APOSPC302990-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :238.14 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1323966-19-3
Formula:C₉H₆F₄O₃
MDL Number:MFCD19687177