4-Methoxy-3-methyl-5-(trifluoromethyl)benzoic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1431329-61-1
:
Formula:C₁₀H₉F₃O₃
MDL Number:MFCD22682822
:234.17 g/mol
Storage Temperature:Ambient
Frequently Bought Together