
4-Methoxy-3-methyl-5-(trifluoromethyl)benzaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1431329-77-9
:
Formula:C₁₀H₉F₃O₂
MDL Number:MFCD22682818
:218.17 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together