
2-Methyl-5-(pentafluorothio)aniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:623943-52-2
:
Formula:C₇H₈F₅NS
MDL Number:MFCD16652447
ADR:6.1,III
:233.20 g/mol
UN:2811
:GHS06
Storage Temperature:Ambient
Frequently Bought Together