
4-Fluoro-3-methyl-5-(trifluoromethyl)benzaldehyde 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1373921-14-2
:
Formula:C₉H₆F₄O
MDL Number:MFCD22201063
:206.14 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together