3-Bromo-5-(trifluoromethoxy)phenylacetic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1092461-35-2
:
Formula:C₉H₆BrF₃O₃
MDL Number:MFCD11519378
:313.07 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together