Order Entry
Italy
ContactUsLinkComponent
3-[2,4-Bis(trifluoromethyl)phenyl]propionic acid 98%
3-[2,4-Bis(trifluoromethyl)phenyl]propionic acid 98%
Catalog # APOSPC302498-1G
Supplier:  Apollo Scientific
CAS Number:  
3-[2,4-Bis(trifluoromethyl)phenyl]propionic acid 98%
Catalog # APOSPC302498-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :286.17 g/mol
Storage Temperature:Ambient
Formula:C₁₁H₈F₆O₂
MDL Number:MFCD11519312