4-Fluoro-3-methyl-5-(trifluoromethyl)phenylacetic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1373920-82-1
:
Formula:C₁₀H₈F₄O₂
MDL Number:MFCD22201074
:236.16 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together