
6-Fluoro-3-methyl-2-(trifluoromethyl)benzaldehyde 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1373920-97-8
:
Formula:C₉H₄F₄O
MDL Number:MFCD22201035
:206.14 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together