4-(Difluoromethoxy)cinnamic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:103962-07-8
:
Formula:C₁₀H₈F₂O₃
MDL Number:MFCD01222822
:214.17 g/mol
Melting Pt:183…186 °C
Storage Temperature:Ambient
Frequently Bought Together