Order Entry
Italy
ContactUsLinkComponent
3-(2-Fluoro-1,1'-biphenyl-4-yl)-1H-pyrazole
3-(2-Fluoro-1,1'-biphenyl-4-yl)-1H-pyrazole
Catalog # APOSPC300993-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(2-Fluoro-1,1'-biphenyl-4-yl)-1H-pyrazole
Catalog # APOSPC300993-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :238.27 g/mol
Storage Temperature:Ambient
CAS Number:1287217-55-3
Formula:C₁₅H₁₁FN₂
MDL Number:MFCD18089275