Order Entry
Italy
ContactUsLinkComponent
Ethyl-4-hydroxy-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate
Irritant
Ethyl-4-hydroxy-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate
Catalog # APOSPC2935-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl-4-hydroxy-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate
Catalog # APOSPC2935-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :317.29 g/mol
Melting Pt:121…123 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:227199-08-8
Formula:C₁₃H₁₀F₃NO₃S
MDL Number:MFCD00816788