Order Entry
Italy
ContactUsLinkComponent
2,5-Difluorophenyl isothiocyanate 98+%
Toxic
2,5-Difluorophenyl isothiocyanate 98+%
Catalog # APOSPC2875M-5G
Supplier:  Apollo Scientific
CAS Number:  
Toxic
2,5-Difluorophenyl isothiocyanate 98+%
Catalog # APOSPC2875M-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :171.17 g/mol
Boiling Pt:112 °C/35mmHg
Density:1.338 g/cm³ (25 °C)
Flash Pt:92 °C
 :GHS06
Storage Temperature:Ambient
CAS Number:206559-57-1
Formula:C₇H₃F₂NS
MDL Number:MFCD00041046