Order Entry
Italy
ContactUsLinkComponent
2,4-Difluoro-DL-phenylalanine 97%
Irritant
2,4-Difluoro-DL-phenylalanine 97%
Catalog # APOSPC2874H-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,4-Difluoro-DL-phenylalanine 97%
Catalog # APOSPC2874H-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
Melting Pt:224…226 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:32133-35-0
Formula:C₉H₉F₂NO₂
MDL Number:MFCD00061302