
(2,4-Difluorophenyl)acetonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:656-35-9
:
Formula:C₈H₅F₂N
MDL Number:MFCD00009971
:153.13 g/mol
Boiling Pt:98 °C/10mmHg
Density:1.249 g/cm³ (25 °C)
Flash Pt:93 °C
Storage Temperature:Ambient
EINECS:211-514-1
UN:3276
ADR:6.1,III
:GHS06
Frequently Bought Together