3-Chloro-N-(2-methoxyethyl)-5-(trifluoromethyl)-2-pyridinamine, Technical Grade
Catalog # APOSPC28045-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:254.64 g/mol
Storage Temperature:Ambient
CAS Number:338406-35-2
Formula:C₉H₁₀ClF₃N₂O
MDL Number:MFCD00172037