Order Entry
Italy
ContactUsLinkComponent
2,4-Difluorobenzylamine 98%
Corrosive
2,4-Difluorobenzylamine 98%
Catalog # APOSPC2735-100G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
2,4-Difluorobenzylamine 98%
Catalog # APOSPC2735-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :143.14 g/mol
Boiling Pt:55…57 °C/8mmHg
Density:1.204 g/cm³ (25 °C)
Flash Pt:68 °C
Storage Temperature:Ambient
EINECS:276-502-0
UN:2735
ADR:8,III
 :GHS05
CAS Number:72235-52-0
Formula:C₇H₇F₂N
MDL Number:MFCD00010142