Order Entry
Italy
ContactUsLinkComponent
4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylic acid 97%
Irritant
4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylic acid 97%
Catalog # APOSPC2605-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylic acid 97%
Catalog # APOSPC2605-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :287.26 g/mol
Melting Pt:237…238 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:144059-86-9
Formula:C₁₂H₈F₃NO₂S
MDL Number:MFCD00068105