Order Entry
Italy
ContactUsLinkComponent
2,6-Dichloro-4-(trifluoromethyl)benzonitrile 97%
Irritant
2,6-Dichloro-4-(trifluoromethyl)benzonitrile 97%
Catalog # APOSPC2554E-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,6-Dichloro-4-(trifluoromethyl)benzonitrile 97%
Catalog # APOSPC2554E-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :240.100 g/mol
Boiling Pt:66 °C/1mmHg
Storage Temperature:Ambient
UN:3276
ADR:6.1,III
 :GHS07
CAS Number:157021-61-9
Formula:C₈H₂Cl₂F₃N
MDL Number:MFCD00220879