Order Entry
Italy
ContactUsLinkComponent
4,5-Dichloro-2-nitrobenzotrifluoride 97%
Irritant
4,5-Dichloro-2-nitrobenzotrifluoride 97%
Catalog # APOSPC2489-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4,5-Dichloro-2-nitrobenzotrifluoride 97%
Catalog # APOSPC2489-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :259.9975 g/mol
Melting Pt:36…38 °C
Density:1.6 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
 :GHS07
CAS Number:50594-31-5
Formula:C₇H₂Cl₂F₃NO₂
MDL Number:MFCD00236666