3-(Difluoromethoxy)phenylacetic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:262587-06-4
:
Formula:C₉H₈F₂O₃
MDL Number:MFCD00236238
Melting Pt:45…51 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together