
2,4-Difluorobenzenesulphonamide
Catalog # APOSPC2353-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:193.17 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:13656-60-5
Formula:C₆H₅F₂NO₂S
MDL Number:MFCD01320743