Order Entry
Italy
ContactUsLinkComponent
 
2-Amino-3,3,3-trifluoropropionic acid hydrochloride 97%
Catalog # APOSPC2052-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Amino-3,3,3-trifluoropropionic acid hydrochloride 97%
Catalog # APOSPC2052-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 3,3,3-Trifluoro-DL-alanine hydrochloride
 :
 :179.53 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:96105-72-5
Formula:C₃H₅ClF₃NO₂
MDL Number:MFCD04973034
Melting Pt:186 °C