4-[4-(Trifluoromethyl)phenoxy]piperidine hydrochloride 95+%
Catalog # APOSPC201221-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:281.7000 g/mol
Storage Temperature:Ambient
CAS Number:287952-09-4
Formula:C₁₂H₁₅ClF₃NO₂
MDL Number:MFCD02258937