
4-[3,5-Di(trifluoromethyl)phenyl]-3-thiosemicarbazide
Catalog # APOSPC2009-2G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:2 g
- Seq:3107
Specifications
About this item
:
:303.23 g/mol
Melting Pt:158…160 °C
:GHS07
Storage Temperature:Ambient
CAS Number:38901-31-4
Formula:C₉H₇F₆N₃S
MDL Number:MFCD00041292