3-Cyano-2-fluorobenzoic acid
Catalog # APOSPC200492-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:165.12 g/mol
Storage Temperature:Ambient
CAS Number:219519-77-4
Formula:C₈H₄FNO₂
MDL Number:MFCD09261249