Order Entry
Italy
ContactUsLinkComponent
3-(4-Fluorophenyl)-1-oxa-3,8-diazaspiro[4.5]decan-2-one
3-(4-Fluorophenyl)-1-oxa-3,8-diazaspiro[4.5]decan-2-one
Catalog # APOSPC200360-500MG
Supplier:  Apollo Scientific
CAS Number:  
3-(4-Fluorophenyl)-1-oxa-3,8-diazaspiro[4.5]decan-2-one
Catalog # APOSPC200360-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :250.28 g/mol
Melting Pt:114…115 °C
Storage Temperature:Ambient
CAS Number:1047655-95-7
Formula:C₁₃H₁₅FN₂O₂
MDL Number:MFCD20232922