5-Methoxy-6-(trifluoromethyl)-1,3-dihydro-2H-indol-2-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1190198-26-5
:
Formula:C₁₀H₈F₃NO₂
MDL Number:MFCD12922678
:231.18 g/mol
Melting Pt:226…228 °C
Storage Temperature:Ambient
Frequently Bought Together