Ethyl-8-iodo-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1135283-39-4
:
Formula:C₁₁H₈F₃IN₂O₂
MDL Number:MFCD11841000
Melting Pt:154…155 °C
Storage Temperature:Ambient
Frequently Bought Together