
Ethyl-3-(trifluoromethyl)quinoxaline-2-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3885-40-3
:
Formula:C₁₂H₉F₃N₂O₂
MDL Number:MFCD08690765
:270.21 g/mol
Melting Pt:47…49 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together