
4-Chloro-3-fluorobenzoic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:403-17-8
:
Formula:C₇H₄ClFO₂
MDL Number:MFCD00143290
:174.56 g/mol
Melting Pt:190…196 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together