
3-Chloro-4-fluorobenzoic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:403-16-7
:
Formula:C₇H₄ClFO₂
MDL Number:MFCD00042477
:174.56 g/mol
Melting Pt:133…136 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together