Order Entry
Italy
ContactUsLinkComponent
1-(3-Trifluoromethylphenyl)-1,2-propandione 95+%
Irritant
1-(3-Trifluoromethylphenyl)-1,2-propandione 95+%
Catalog # APOSPC1661-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-(3-Trifluoromethylphenyl)-1,2-propandione 95+%
Catalog # APOSPC1661-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :216.16 g/mol
Boiling Pt:80 °C/0.07mmHg
Melting Pt:25…26 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:10557-15-0
Formula:C₁₀H₇F₃O₂
MDL Number:MFCD01941304