
2-Methylsulphonyl-1-(2-trifluoromethylphenyl)ethanone 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:387350-34-7
:
Formula:C₁₀H₉F₃O₃S
MDL Number:MFCD01941307
:266.24 g/mol
Melting Pt:61…63 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together