


tert-Butyldimethylsilyl trifluoromethanesulphonate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:69739-34-0
:
Formula:C₇H₁₅F₃O₃SSi
MDL Number:MFCD00000405
:264.34 g/mol
Boiling Pt:65…67 °C/12mmHg
Density:1.151 g/cm³ (25 °C)
Flash Pt:36 °C
EINECS:274-102-0
UN:2920
ADR:8+3,II
:GHS02
:GHS05
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together