
3-Bromo-α,α,α,5-tetrafluorotoluene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:130723-13-6
:
Formula:C₇H₃BrF₄
MDL Number:MFCD00042498
:243 g/mol
Boiling Pt:146 °C
Density:1.511 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together