2-Bromo-4,5-difluorophenol 98%
Codice catalogo
Product Details & Documents
Synonyms: 1-Bromo-3,4-difluoro-6-hydroxybenzene
- CAS Number:166281-37-4
- Formula:C₆H₃BrF₂O
- MDL Number:MFCD00070751
- Molecular Weight:208.99 g/mol
- Density:1.829 g/cm³ (25 °C)
- Flash Pt:81 °C
- Storage Temperature:Ambient



