Order Entry
Italy
ContactUsLinkComponent
trans-1,2-Bis(perfluoro-n-butyl)ethylene 99%
Irritant
trans-1,2-Bis(perfluoro-n-butyl)ethylene 99%
Catalog # APOSPC1237-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
trans-1,2-Bis(perfluoro-n-butyl)ethylene 99%
Catalog # APOSPC1237-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Boiling Pt:64 °C/56mmHg
Density:1.675 g/cm³ (25 °C)
 :464.1 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:84551-43-9
Formula:C₁₀H₂F₁₈
MDL Number:MFCD00055550