Order Entry
Italy
ContactUsLinkComponent
Barium fluoride 99%
Toxic
Barium fluoride 99%
Catalog # APOSPC1170-25G
Supplier:  Apollo Scientific
CAS Number:  
Toxic
Barium fluoride 99%
Catalog # APOSPC1170-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Boiling Pt:2260 °C
Melting Pt:1353 °C
Density:4.89 g/cm³ (25 °C)
Storage Temperature:Ambient
ADR:6.1,III
 :175.33 g/mol
EINECS:232-108-0
UN:1564
 :GHS06
CAS Number:7787-32-8
Formula:BaF₂
MDL Number:MFCD00003450