
3,5-Difluoro-2-methoxybenzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:886498-75-5
:
Formula:C₈H₆F₂O₃
MDL Number:MFCD06660198
:188.13 g/mol
Melting Pt:98…103 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together