

Methyl-2,4-difluorobenzoate 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:106614-28-2
:
Formula:C₈H₆F₂O₂
MDL Number:MFCD03093799
:172.13 g/mol
Boiling Pt:128…129 °C
Density:1.31 g/cm³ (25 °C)
Flash Pt:82 °C
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together