Order Entry
Italy
ContactUsLinkComponent
1-[3-(Trifluoromethyl)phenyl]-1H-pyrrole-2,5-dione
1-[3-(Trifluoromethyl)phenyl]-1H-pyrrole-2,5-dione
Catalog # APOSPC11122-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[3-(Trifluoromethyl)phenyl]-1H-pyrrole-2,5-dione
Catalog # APOSPC11122-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :241.17 g/mol
Storage Temperature:Ambient
CAS Number:53629-19-9
Formula:C₁₁H₆F₃NO₂
MDL Number:MFCD00439448