
2-Bromo-4-(trifluoromethyl)benzaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:85118-24-7
:
Formula:C₈H₄BrF₃O
MDL Number:MFCD09835101
Boiling Pt:229 °C
Flash Pt:92 °C
:253.0200 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together