
3-Bromo-4-(trifluoromethoxy)benzaldehyde 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:85366-66-1
:
Formula:C₈H₄BrF₃O₂
MDL Number:MFCD08458004
:269.0200 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together