Order Entry
Italy
ContactUsLinkComponent
Ammonium hydrogen difluoride 98%
Ammonium hydrogen difluoride 98%
Catalog # APOSPC1104-500G
Supplier:  Apollo Scientific
CAS Number:  
Ammonium hydrogen difluoride 98%
Catalog # APOSPC1104-500G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :57.04 g/mol
Boiling Pt:230 °C/760mmHg
Melting Pt:127 °C
Density:1.5 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:1727
ADR:8,II
CAS Number:1341-49-7
Formula:NH₄HF₂
MDL Number:MFCD00012018