(Perfluoro-n-octyl)phenyliodonium trifluoromethanesulphonate
Catalog # APOSPC10692-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:772.13 g/mol
Storage Temperature:Ambient
CAS Number:77758-89-5
Formula:C₁₅H₅F₂₀IO₃S
MDL Number:MFCD00135120